missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACE/ADAM17 Antibody (136133), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB21291D
This item is not returnable.
View return policy
Description
TACE/ADAM17 Monoclonal antibody specifically detects TACE/ADAM17 in Human samples. It is validated for Western Blot, Flow Cytometry, KnockDown
Specifications
| TACE/ADAM17 | |
| Monoclonal | |
| DyLight 350 | |
| ADAM 17, ADAM metallopeptidase domain 17, ADAM metallopeptidase domain 18, ADAM18, CD156B, CD156b antigen, CSVP, disintegrin and metalloproteinase domain-containing protein 17, EC 3.4.24.86, MGC71942, Snake venom-like protease, TACEcSVP, TNF-alpha convertase, TNF-alpha converting enzyme, TNF-alpha-converting enzyme, tumor necrosis factor, alpha, converting enzyme | |
| E. coli-derived recombinant human TACE/ADAM17, Asp695-Cys824, Accession # P78536 | |
| 0.5 mL | |
| Alzheimers Research, Apoptosis, Myeloid derived Suppressor Cell, Neurodegeneration, Stem Cell Signaling Pathway | |
| 6868 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, KnockDown | |
| 136133 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction