missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TACI/TNFRSF13B/CVID Antibody (166010), PE/Cy5.5, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1041PECY55
This item is not returnable.
View return policy
Description
TACI/TNFRSF13B/CVID Monoclonal antibody specifically detects TACI/TNFRSF13B/CVID in Mouse samples. It is validated for Flow Cytometry
Specifications
| TACI/TNFRSF13B/CVID | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 166010 | |
| Flow Cytometry | |
| CD267, CD267 antigen, CVID, CVID2, FLJ39942, MGC133214, TACIMGC39952, TNFRSF14B, Transmembrane activator and CAML interactor, tumor necrosis factor receptor 13B, tumor necrosis factor receptor superfamily member 13B, tumor necrosis factor receptor superfamily, member 13B | |
| Mouse myeloma cell line NS0-derived recombinant mouse TACI/TNFRSF13B, Phe5-Thr129, Accession # Q9ET35 | |
| 0.5 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Signal Transduction | |
| 23495 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction