missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TACR1 Recombinant Rabbit Monoclonal Antibody (K01_1U72)
GREENER_CHOICE

Product Code. p-200114345
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826148 100 μL
30824779 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826148 Supplier Invitrogen™ Supplier No. MA563089

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

TACR1 Recombinant Monoclonal Antibody for Western Blot

Tachykinin Receptor 1, a member of the Tachykinin Receptor subfamily, is involved in the mediation of phosphatidylinositol metabolism by the tachykinin substance P, also referred to as neurokinin 1. It is associated with the management of depression, anxiety, and stress. It is also involved in regulating behavioral responses, neuronal survival, and cardiovascular and respiratory functions. The tachykinin receptor 1 is implicated in the progression of rheumatoid arthritis and osteoarthritis. Two variants of the tachykinin receptor 1 are produced by alternative splicing. Tachykinin receptor 1 has been reported primarily in brain, and in blood lymphocytes, nose, small intestine, and stomach. ESTs have been isolated from brain and spleen libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TACR1
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1U72
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Tacr1
Gene Accession No. P25103, P30548
Gene Alias neurokinin 1 receptor; neurokinin receptor 1; neurokinin receptor subtype NK1; neurokinin-1 receptor; Nk1 receptor; NK-1 receptor; Nk1r; NK-1R; NKIR; SPR; substance p receptor; substance-P receptor; TAC1R; tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor); TACR1; truncated neurokinin-1 receptor
Gene Symbols Tacr1
Host Species Rabbit
Immunogen A synthetic peptide of human Neurokinin 1 Receptor.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21336, 6869
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.