missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TACR2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA533071

Product Code. 13474739

  • € 520.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Pig (100%); Rat, Bovine, Dog, Hamster, Elephant, Panda, Horse, Rabbit, Guinea pig (95%); Monkey, Marmoset, Mouse, Bat (89%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

This gene belongs to a family of genes that function as receptors for tachykinins. Receptor affinities are specified by variations in the 5'-end of the sequence. The receptors belonging to this family are characterized by interactions with G proteins and 7 hydrophobic transmembrane regions. This gene encodes the receptor for the tachykinin neuropeptide substance K, also referred to as neurokinin A.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TACR2
Polyclonal
Unconjugated
TACR2
7 transmembrane receptor; neurokinin 2 receptor; neurokinin A receptor; NK2; NK-2 receptor; NK2R; NK-2R; NKNAR; seven transmembrane helix receptor; SK receptor; SKR; substance K receptor; substance-K receptor; TAC2R; Tachykinin 2 (substance K) receptor; tachykinin NK-2 receptor; tachykinin receptor 2; TACR2
Rabbit
Antigen affinity chromatography
RUO
100151877, 6865
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
P21452
TACR2
Synthetic 19 amino acid peptide from C-terminus of human Neurokinin A Receptor.
50 μg
Primary
Human, Pig
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.