missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TADA1L Antibody (PCRP-TADA1-1C9) - Azide and BSA Free, Novus Biologicals™
Shop All Bio Techne ProductsDescription
TADA1L Monoclonal antibody specifically detects TADA1L in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Protein Array
Specifications
Specifications
| Antigen | TADA1L |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Peptide Array |
| Classification | Monoclonal |
| Clone | PCRP-TADA1-1C9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunoprecipitation 1-2 ug per 100-500 ug protein, Protein Array |
| Formulation | 10mM PBS |
| Gene Alias | ADA1, hADA1, HFI1, SPT3-associated factor 42, SPT3-associated factor 42 (STAF42), STAF42transcriptional adaptor 1 (HFI1 homolog, yeast)-like, TADA1LRP1-9E21.4, transcriptional adapter 1, Transcriptional adapter 1-like protein, transcriptional adaptor 1, transcriptional adaptor 1 (HFI1 homolog, yeast) |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?