missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TADA2L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104485
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ADA2L.
TADA2L (transcriptional adapter 2-like), also known as TADA2A (transcriptional adapter 2-alpha) or ADA2-like protein, is a 443 amino acid nuclear protein that exists as 2 alternatively spliced isoforms. While most abundantly expressed in testis, TADA2L is present in all tissues. TADA2L contains one SANT domain and one SWIRM domain, and interacts with GCN5 and GR (NR3C1). Its ability to bind double-stranded DNA allows TADA2L to play a role in chromatin remodeling. Although it makes up part of the PCAF complex, TADA2L is also a component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4. The gene that encodes TADA2L contains 71,408 bases and maps to human chromosome 17q12. Chromosome 7 houses over 1,000 genes, comprises nearly 5% of the human genome and has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman-Diamond syndrome.
Specifications
| TADA2L | |
| Polyclonal | |
| Unconjugated | |
| TADA2A | |
| ADA2; ADA2A; ADA2-like protein; AV319371; D030022J10Rik; hADA2; KL04P; TADA2A; TADA2L; transcriptional adapter 2-alpha; Transcriptional adapter 2-like; transcriptional adaptor 2 alpha; transcriptional adaptor 2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 217031, 360581, 6871 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75478, Q6AYE3, Q8CHV6 | |
| TADA2A | |
| A synthesized peptide derived from human TADA2A(Accession O75478), corresponding to amino acid residues R218-K268. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction