missing translation for 'onlineSavingsMsg'
Learn More

TADA2L Polyclonal Antibody, Invitrogen™

Product Code. 15515471
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515471 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15515471 Supplier Invitrogen Supplier No. PA538547

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TADA2L (transcriptional adapter 2-like), also known as TADA2A (transcriptional adapter 2-alpha) or ADA2-like protein, is a 443 amino acid nuclear protein that exists as 2 alternatively spliced isoforms. While most abundantly expressed in testis, TADA2L is present in all tissues. TADA2L contains one SANT domain and one SWIRM domain, and interacts with GCN5 and GR (NR3C1). Its ability to bind double-stranded DNA allows TADA2L to play a role in chromatin remodeling. Although it makes up part of the PCAF complex, TADA2L is also a component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4. The gene that encodes TADA2L contains 71,408 bases and maps to human chromosome 17q12. Chromosome 7 houses over 1,000 genes, comprises nearly 5% of the human genome and has been linked to osteogenesis imperfecta, Pendred syndrome, lissencephaly, citrullinemia and Shwachman-Diamond syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TADA2L
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TADA2A
Gene Accession No. O75478, Q8CHV6, Q6AYE3
Gene Alias Transcriptional adapter 2-alpha, Transcriptional adapter 2-like, ADA2-like protein, TADA2A, TADA2L, KL04P
Gene Symbols TADA2A
Host Species Rabbit
Immunogen A synthetic peptide derived from the internal region of human ADA2L
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217031, 360581, 6871
Target Species Human, Mouse, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.