Learn More
Melford TAE 50X
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Solution contains:
Tris (hydroxymethyl) aminomethane: 2 M
Acetic Acid: 1 M
EDTA, Disodium Salt Dihydrate: 50 mM
Specifications
Specifications
| CAS | 77-86-1, 64-19-7, 6381-92-6, 7732-18-5 |
| Quantity | 1 L |