missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1 48 (TAF1A) Monoclonal Antibody (OTI8C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF811373
This item is not returnable.
View return policy
Description
TAF1 48 (TAF1A) Monoclonal antibody specifically detects TAF1 48 (TAF1A) in Human samples. It is validated for Western BlotSpecifications
| TAF1 48 (TAF1A) | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P36959 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2766 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI8C10 | |
| Unconjugated | |
| GMPR | |
| MGC:17061; mTAFI48; RAFI48; RNA polymerase I-specific TBP-associated factor 48 kDa; similar to TBP-associated factor 1A isoform 1; SL1; SL1, 48kD subunit; Taf1a; TAFI48; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, A; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; TATA box binding protein associated factor 1A; TATA box binding protein-associated factor TAF1A; TATA box-binding protein-associated factor 1A; TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I, A; TBP-associated factor 1A; Transcription factor SL1; transcription initiation factor SL1/TIF-IB subunit A | |
| Full length human recombinant protein of human GMPR produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction