missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF13 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | TAF13 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TAF13 Polyclonal antibody specifically detects TAF13 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| TAF13 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 18kDa, MGC22425, TAF(II)18, TAF13 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2K, TAFII18TAFII-18, TATA box binding protein (TBP)-associated factor, RNA polymerase II, K, 18kD, transcription initiation factor TFIID 18 kD subunit, Transcription initiation factor TFIID 18 kDa subunit, transcription initiation factor TFIID subunit 13 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-124 of human TAF13 (NP_005636.1). MADEEEDPTFEEENEEIGGGAEGGQGKRKRLFSKELRCMMYGFGDDQNPYTESVDILEDLVIEFITEMTHKAMSIGRQGRVQVEDIVFLIRKDPRKFARVKDLLTMNEELKRARKAFDEANYGS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 6884 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title