missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF15 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-568G
This item is not returnable.
View return policy
Description
TAF15 Polyclonal antibody specifically detects TAF15 in Human samples. It is validated for Western Blot
Specifications
| TAF15 | |
| Polyclonal | |
| Western Blot | |
| 68kDa, hTAFII68, Npl3, RBP56TAFII68, RNA-binding protein 56, TAF(II)68, TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2NRBP56/CSMF fusion, TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD(RNA-binding protein 56), TATA box-binding protein-associated factor 2N (RNA-binding protein 56), TATA-binding protein-associated factor 2N, TBP-associated factor 15,68 kDa TATA-binding protein-associated factor | |
| A synthetic peptide which maps to a region between residues 550 and the C-terminus (residue 592) of human TAFII68 using the numbering given in Swiss-Prot entry Q92804 (GeneID 8148). | |
| 0.1 mL | |
| DNA Repair, Neuroscience | |
| 8148 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction