missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1A Polyclonal Antibody

TAF1A Polyclonal Antibody
Brand: Invitrogen PA5101172
This item is not returnable.
View return policy
Description
TAF1A Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| TAF1A | |
| Polyclonal | |
| Unconjugated | |
| TAF1A | |
| MGC:17061; mTAFI48; RAFI48; RNA polymerase I-specific TBP-associated factor 48 kDa; similar to TBP-associated factor 1A isoform 1; SL1; SL1, 48kD subunit; Taf1a; TAFI48; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, A; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; TATA box binding protein associated factor 1A; TATA box binding protein-associated factor TAF1A; TATA box-binding protein-associated factor 1A; TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I, A; TBP-associated factor 1A; Transcription factor SL1; transcription initiation factor SL1/TIF-IB subunit A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21339, 360893, 9015 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97357, Q15573, Q3B7U2 | |
| TAF1A | |
| A synthesized peptide derived from human TAF1A(Accession Q15573), corresponding to amino acid residues K298-A348. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction