missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1B Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808684S
This item is not returnable.
View return policy
Description
TAF1B Monoclonal antibody specifically detects TAF1B in Human samples. It is validated for Western BlotSpecifications
| TAF1B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6UXH0 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55908 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| ANGPTL8 | |
| 4930408G01Rik; A230108M10Rik; MGC:9349; p63; RAF1B; RAFI63; RNA polymerase I-specific TBP-associated factor 63 kDa; RNA polymerase I-specific TBP-associated factor 68 kDa; SL1; SL1, 63kD subunit; Taf1b; TAFI63; TAFI68; Tafi86; TATA box binding protein (Tbp)-associated factor, RNA polymerase I, B; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, B, 63kDa; TATA box binding protein associated factor 1B; TATA box-binding protein-associated factor 1B; TATA box-binding protein-associated factor RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I subunit B; TATA-box binding protein associated factor, RNA polymerase I, B; TBP-associated factor 1B; TBP-associated factor RNA polymerase I 68; transcription factor TAFI68; transcription initiation factor SL1/TIF-IB subunit B | |
| Human recombinant protein fragment corresponding to amino acids 22-198 of human C19ORF80 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction