missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38327-100ul
This item is not returnable.
View return policy
Description
TAF1C Polyclonal antibody specifically detects TAF1C in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TAF1C | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| MGC:39976, RNA polymerase I-specific TBP-associated factor 110 kDa, SL1, 110kD subunit, SL1TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kD, TAFI110TBP-associated factor 1C, TAFI95, TATA box binding protein (TBP)-associated factor, RNA polymerase I, C, 110kDa, TATA box-binding protein-associated factor 1C, TATA box-binding protein-associated factor RNA polymerase I subunit C, transcription factor SL1, Transcription initiation factor SL1/TIF-IB subunit C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 70-320 of human TAF1C (NP_001230085.1).,, Sequence:, PPLIDPWDPGLTARDLLFRGGYRYRKRPRVVLDVTEQISRFLLDHGDVAFAPLGKLMLENFKLEGAGSRTKKKTVVSVKKLLQDLGGHQPWGCPWAYLSNRQRRFSILGGPILGTSVASHLAELLHEELVLRWEQLLLDEACTGGALAWVPGRTPQFGQLVYPAGGAQDRLHFQEVVLTPGDNPQFLGKPGRIQLQGPVRQVVTCTVQGETLLAVRSDYHCAVWKFGKQWQPTLLQAMQVEKGATGISLSP | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9013 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction