missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF2 Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-21372AF700
This item is not returnable.
View return policy
Description
TAF2 Polyclonal antibody specifically detects TAF2 in Human samples. It is validated for Western Blot
Specifications
| TAF2 | |
| Polyclonal | |
| Western Blot | |
| 150 kDa cofactor of initiator function, CIF150TAFII-150, cofactor of initiator function, 150kD subunit, TAF(II)150, TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF2B, TAFII150RNA polymerase II TBP-associated factor subunit B, TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD, TBP-associated factor 150 kDa, Transcription initiation factor TFIID 150 kDa subunit, transcription initiation factor TFIID subunit 2 | |
| The immunogen recognized by this antibody maps to a region between residue 1149 and 1199 of human TBP-associated factor 2 using the numbering given in entry NP_003175.1 (GeneID 6873). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 6873 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction