Learn More
TAF2 Monoclonal Antibody (OTI2A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504588S
Description
TAF2 Monoclonal antibody specifically detects TAF2 in Human samples. It is validated for Western BlotSpécification
| TAF2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48742 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3975 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2A5 | |
| Unconjugated | |
| Lhx1 | |
| 150 kDa cofactor of initiator function; 150kDa; 4732460C16Rik; AI425886; CIF150; cofactor of initiator function, 150kD subunit; fc30f02; fc56a05; MRT40; RNA polymerase II TBP-associated factor subunit B; TAF(II)150; TAF150; TAF2; taf2 protein; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF2 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 150kDa; TAF2 RNA polymerase II, TATA box binding protein-associated factor, 150 kD; TAF2B; Taf2p; TAFII150; TAFII-150; TAFIIB; TATA box binding protein (TBP)-associated factor 150kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, B, 150kD; TATA box binding protein associated factor 2; TATA-box binding protein associated factor 2; TBP-associated factor 150 kDa; TBP-associated factor 2; transcription initiation factor TFIID 150 kDa subunit; Transcription initiation factor TFIID subunit 2; transcription initiation factor TFIID subunit 2-like protein; TSM1; TSM-1; wu:fc30f02; wu:fc56a05; YCR042C; YCR42C; YCR724 | |
| Human recombinant protein fragment corresponding to amino acids 100-362 of human LHX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Le programme Encompass de Fisher Scientific propose des articles qui ne font pas partie de notre portefeuille de distribution. Ces produits n'ont généralement pas de photos ou de descriptions détaillées. Cependant, nous nous engageons à améliorer votre expérience d'achat. Veuillez utiliser le formulaire ci-dessous pour nous faire part de vos commentaires sur le contenu de ce produit.