missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF3 Antibody (S01-6A4), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 421.00 - € 645.00
Specifications
| Antigen | TAF3 |
|---|---|
| Clone | S01-6A4 |
| Concentration | 0.3 mg/mL |
| Dilution | Western Blot, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18612484
|
Novus Biologicals
NBP3-19497-100ul |
100 μg |
€ 645.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18621884
|
Novus Biologicals
NBP3-19497-50ul |
50 μg |
€ 421.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAF3 Monoclonal antibody specifically detects TAF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TAF3 | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| 140 kDa TATA box-binding protein-associated factor, MGC25063, RNA polymerase II transcription factor TAFII140, TAF(II)140, TAF140, TAF3 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 140kDa, TAFII140, TAFII-140, TBP-associated factor 3, transcription initiation factor TFIID 140 kDa subunit | |
| Recombinant protein of human TAF3 (Uniprot # Q5VWG9) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S01-6A4 | |
| Western Blot, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 83860 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title