missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Taf4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80705
This item is not returnable.
View return policy
Description
Taf4 Polyclonal specifically detects Taf4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Taf4 | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| O00268 | |
| TAF4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VLVRSENGQLLMIPQQALAQMQAQAHAQPQTTMAPRPATPTSAPPVQISTVQAPGTPIIARQVTPTTIIKQVSQAQTTVQPSATLQRSPGVQPQLVLGGAAQTASLGTATAVQTGTPQRTVPGATTTSSAATETMENVKKCK | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FLJ41943, TAF(II)130, TAF(II)135, TAF2C1TBP-associated factor 4, TAF2CRNA polymerase II TBP-associated factor subunit C, TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF4A, TAFII130TAFII-130, TAFII135TAFII-135, TATA box binding protein (TBP)-associated factor, 135kD, TATA box binding protein (TBP)-associated factor, RNA polymerase II, C1, 130kD, Transcription initiation factor TFIID 130 kDa subunit, transcription initiation factor TFIID 135 kD subunit, transcription initiation factor TFIID subunit 4 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6874 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction