missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16660925 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16660925 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16660925 Supplier Invitrogen™ Supplier No. PA5104475

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAF4.

TAF II p105, also called TAF4B, is a celltype specific transcriptional co-activator that is a component of the TFIID complex. Expressed primarily in B cells and ovarian granulosa cells, TAF II p105 can interact with OCBA/POU2AF1 to activate B cell-specific octamerdependent transcription. Additionally, TAF II p105 plays an important role in co-activating the transcription factor NFkB and is essential for activation of anti-apoptotic genes such as TNFAIP3. TAF II p135, also known as TAF4, TAF2C, TAF2C1, TAF4A or TAFII130, is a 1,085 amino acid subunit of TFIID that accelerates transcriptional activation triggered by thyroid hormone (TR) or retinoic acid (RA). Localized to the nucleus, TAF II p135 contains one TAFH domain and is thought to bind to proteins that contain glutamine-rich domains, such as the transcription factor CREB.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAF4
Gene Accession No. O00268
Gene Alias 135kDa; AI450312; FLJ41943; LOW QUALITY PROTEIN: transcription initiation factor TFIID subunit 4; RNA polymerase II TBP-associated factor subunit C; TAF(II)130; TAF(II)135; TAF2C; TAF2C1; TAF4; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135kDa; Taf4a; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor, 135 kD; TAFII 130; TAFII 135; TAFII130; TAFII-130; TAFII135; TAFII-135; TATA box binding protein (TBP)-associated factor, RNA polymerase II, C1, 130kD; TATA box binding protein associated factor 4; TATA-box binding protein associated factor 4; TBP-associated factor 4; Transcription initiation factor TFIID 130 kDa subunit; transcription initiation factor TFIID 135 kD subunit; transcription initiation factor TFIID 135 kDa subunit; Transcription initiation factor TFIID subunit 4
Gene Symbols TAF4
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAF4(Accession O00268), corresponding to amino acid residues R957-T1007.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228980, 682097, 6874
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.