missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TAF7L Polyclonal Antibody
GREENER_CHOICE

Product Code. 16617244
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16617244 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16617244 Supplier Invitrogen™ Supplier No. PA5100758

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TAF7L.

Probably functions as a spermatogensis-specific component of the DNA-binding general transcription factor complex TFIID, a multimeric protein complex that plays a central role in mediating promoter responses to various activators and repressors. May play a role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TAF7L
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TAF7L
Gene Accession No. Q5H9L4, Q9D3R9
Gene Alias 4933438I11Rik; 50kDa; cancer/testis antigen 40; CT40; RGD1564898; RNA polymerase II TBP-associated factor subunit Q; TAF2Q; Taf7l; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q; TATA box binding protein associated factor 7 like; TATA box binding protein-associated factor RNA polymerase II subunit Q; TATA box-binding protein-associated factor 50 kDa; TATA-box binding protein associated factor 7 like; TBP-associated factor, RNA polymerase II, Q; transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like
Gene Symbols TAF7L
Host Species Rabbit
Immunogen A synthesized peptide derived from human TAF7L(Accession Q5H9L4), corresponding to amino acid residues Q409-F459.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54457, 74469
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.