missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF7L Polyclonal Antibody

TAF7L Polyclonal Antibody
Brand: Invitrogen PA5142502
This item is not returnable.
View return policy
Description
TAF7L Polyclonal Antibody for Western Blot
Specifications
| TAF7L | |
| Polyclonal | |
| Unconjugated | |
| TAF7L | |
| 4933438I11Rik; 50kDa; cancer/testis antigen 40; CT40; RGD1564898; RNA polymerase II TBP-associated factor subunit Q; TAF2Q; Taf7l; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF7-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, 50kDa; TATA box binding protein (TBP)-associated factor, RNA polymerase II, Q; TATA box binding protein associated factor 7 like; TATA box binding protein-associated factor RNA polymerase II subunit Q; TATA box-binding protein-associated factor 50 kDa; TATA-box binding protein associated factor 7 like; TBP-associated factor, RNA polymerase II, Q; transcription initiation factor TFIID 50 kDa subunit; Transcription initiation factor TFIID subunit 7-like | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 54457 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5H9L4 | |
| TAF7L | |
| Peptide with sequence C-NHFQSVLEQLELQEK (aa 429-443). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion