missing translation for 'onlineSavingsMsg'
Learn More

TAF9b Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 196.00 - € 468.00

Specifications

Antigen TAF9b
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18633662
View Documents
Novus Biologicals
NBP2-94263-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18633502
View Documents
Novus Biologicals
NBP2-94263-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

TAF9b Polyclonal antibody specifically detects TAF9b in Mouse samples. It is validated for Western Blot
Specifications

Specifications

TAF9b
Western Blot
Unconjugated
Rabbit
Mouse
31kDa, DN7, DN-7Transcription-associated factor TAFII31L, Neuronal cell death-related protein 7, TAF9B RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF9-like RNA polymerase II, TATA box binding protein (TBP)-associated factor, TAF9Ltranscription associated factor TAFII31L, TAFII31LTBP-associated factor 9L, TFIID-31, transcription initiation factor IID, 31kD subunit, transcription initiation factor TFIID subunit 9B, Transcription initiation factor TFIID subunit 9-like
A synthetic peptide corresponding to a sequence within amino acids 50-150 of human TAF9B (NP_057059.2). ILDDAKIYSSHAKKPNVDADDVRLAIQCRADQSFTSPPPRDFLLDIARQKNQTPLPLIKPYAGPRLPPDRYCLTAPNYRLKSLIKKGPNQGRLVPRLSVGA
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:500-1:2000
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
51616
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.