missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAFA3/FAM19A3 Antibody (460904), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4677N
This item is not returnable.
View return policy
Description
TAFA3/FAM19A3 Monoclonal antibody specifically detects TAFA3/FAM19A3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| TAFA3/FAM19A3 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 284467 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, CyTOF | |
| 460904 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| Chemokine-like protein TAFA-3, family with sequence similarity 19 (chemokine (C-C motif)-like), member A3, MGC138473, TAFA3, TAFA-3 | |
| E. coli-derived recombinant human TAFA3/FAM19A3, Ala31-Arg133, Accession # Q7Z5A8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction