missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TAFA4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106979
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
This gene is a member of the TAFA family which is composed of five highly homologous genes that encode small secreted proteins. These proteins contain conserved cysteine residues at fixed positions, and are distantly related to MIP-1alpha, a member of the CC-chemokine family. The TAFA proteins are predominantly expressed in specific regions of the brain, and are postulated to function as brain-specific chemokines or neurokines, that act as regulators of immune and nervous cells. Alternatively spliced transcript variants have been observed for this gene.
Specifications
| TAFA4 | |
| Polyclonal | |
| Unconjugated | |
| FAM19A4 | |
| C130034I18Rik; Chemokine-like protein TAFA-4; chemokine-like protein TAFA-4; protein FAM19A4; FAM19A4; Fam19a4 Tafa-4; family with sequence similarity 19 (chemokine (C-C motif)-like), member A4; family with sequence similarity 19 member A4, C-C motif chemokine like; family with sequence similarity 19, member A4; protein FAM19A4; TAFA chemokine like family member 4; TAFA4; Tafa-4; TAFA4 protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 151647 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q96LR4 | |
| FAM19A4 | |
| Synthetic peptide directed towards the middle region of human FAM19A4 (aa 35-85). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction