missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tafazzin/TAZ Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98750B
This item is not returnable.
View return policy
Description
Tafazzin/TAZ Polyclonal specifically detects Tafazzin/TAZ in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Tafazzin/TAZ | |
| Polyclonal | |
| PBS | |
| TAFAZZIN | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Tafazzin/TAZ. | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 6901 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| BTHSEFE, cardiomyopathy, dilated 3A (X-linked), CMD3A, EFE2FLJ27390, G4.5endocardial fibroelastosis 2, LVNCX, Protein G4.5, tafazzin, Taz1, XAP-2 | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction