missing translation for 'onlineSavingsMsg'
Learn More

TAGLN Mouse anti-Human, Clone: TAGLN/247, Abnova™

Product Code. 16012926
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16012926 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16012926 Supplier Abnova Supplier No. MAB14610.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against full length recombinant human TAGLN.

  • High specificity
  • High reactivity

Specifications

Antigen TAGLN
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone TAGLN/247
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant human TAGLN.
Dilution Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10 mM PBS.
Gene Accession No. Q01995
Gene Alias DKFZp686P11128/SM22/SMCC/TAGLN1/WS3-10
Gene Symbols TAGLN
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human TAGLN.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6876
Test Specificity This antibody recognizes a 22 kDa protein, identified as TAGLN, also designated SM22-alpha. It may cross-react with SM22-beta.
Target Species Human
Content And Storage Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.