missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TAK 715

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Inhibitor of p38 MAPK (IC50 = 7.1 nM for p38 MAPKα). Also inhibits Wnt-3a-stimulated β-catenin signaling. Inhibits 22 kinases by more than 80%, including CK1δ/ε. Anti-inflammatory and anti-rheumatoid arthritis agent. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 399.51 |
| Purity | >99% |
| Molecular Weight (g/mol) | 399.512 |
| Chemical Name or Material | TAK 715 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction