missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK-960 dihydrochloride, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
TAK-960 dihydrochloride is an orally available, selective inhibitor of polo-like kinase 1 (PLK1), with an IC50 of 0.8 nM. TAK-960 dihydrochloride also shows inhibitory activities against PLK2 and PLK3, with IC50s of 16.9 and 50.2 nM, respectively. TAK-960 dihydrochloride inhibits proliferation of multiple cancer cell lines and exhibits significant efficacy against multiple tumor xenografts.
Specifications
Specifications
| Chemical Name or Material | TAK-960 dihydrochloride |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C27H36Cl2F3N7O3 |
| Quantity | 100 mg |
| Solubility Information | H2O : 10 mg/mL (15.76 mM; Need ultrasonic) ∣DMSO : 3.23 mg/mL (5.09 mM; Need ultrasonic) |
| SMILES | [H]Cl.O=C(NC1CCN(C)CC1)C2=CC(OC)=C(NC3=NC=C(N4C)C(N(C5CCCC5)CC(F)(F)C4=O)=N3)C=C2F.[H]Cl |
| Molecular Weight (g/mol) | 634.52 |
| Formula Weight | 634.52 |
| Percent Purity | 99.81% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction