missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAK1, p Ser439 Antibody (SR2176), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21597-50ul
This item is not returnable.
View return policy
Description
TAK1 Monoclonal antibody specifically detects TAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TAK1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR2176 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| EC 2.7.11, EC 2.7.11.25, MEKK7, mitogen-activated protein kinase kinase kinase 7, TAK1/MAP3K7, TAK1TGF1a, TGF-beta activated kinase 1, TGF-beta-activated kinase 1, Transforming growth factor-beta-activated kinase 1 | |
| A synthesized peptide derived from human phospho-TAK1 [Ser439] (Uniprot #: O43318) | |
| 50 μg | |
| Apoptosis, Immunology, Innate Immunity, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 6885 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction