missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals TAK1L Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00056911-B01P
This item is not returnable.
View return policy
Description
TAK1L Polyclonal antibody specifically detects TAK1L in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| TAK1L | |
| Polyclonal | |
| Western Blot 1:500 | |
| NP_064537.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 56911 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| PBS (pH 7.4) | |
| chromosome 21 open reading frame 7, HC21ORF7, putative gene, TGF-beta-activated kinase like10TAK1LTAK1-like protein, TAK1-like protein 1, TAK1-like protein 2, TAK1-like protein 4, TAKL, TAKL-1, TAKL-2, TAKL-4, TGF-beta activated kinase | |
| C21orf7 (NP_064537.1, 1 a.a. - 242 a.a.) full-length human protein. MVQLIAPLEVMWNEAADLKPLALSRRLECSGGIMAHYSPDLLGPEMESRYFAQVGLEHLASSSPPAFGFLKCLDYSISVLCSATSLAMLEDNPKVSKLATGDWMLTLKPKSITVPVEIPSSPLDDTPPEDSIPLVFPELDQQLQPLPPCHDSEESMEVFKQHCQIAEEYHEVKKEITLLEQRKKELIAKLDQAEKEKVDAAELVREFEALTEENRTLRLAQSQCVEQLEKLRIQYQKRQGSS | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction