Learn More
TAL1 Monoclonal Antibody (OTI3E2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503899
This item is not returnable.
View return policy
Description
TAL-1 is a basic helix-loop-helix transcription. It regulates differentiation and survival during hemopoiesis. TAL-1 is also implicated in the genesis of hemopoietic maligcies and it may play an important role in hemopoietic differentiation. It serves as a positive regulator of eryhtroid differentiation. Mutations are associated with T-cell leukemia and melanoma. TAL-1 is know to bind to the LIM domain containing protein Rhombotin-2.Specifications
| TAL1 | |
| Monoclonal | |
| 0.94 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17542 | |
| TAL1 | |
| Full length human recombit protein of human TAL1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3E2 | |
| Unconjugated | |
| TAL1 | |
| SCL, TCL5, bHLHa17, tal-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6886 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |