missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAL1 Monoclonal Antibody (OTI6H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503919
This item is not returnable.
View return policy
Description
TAL-1 is a basic helix-loop-helix transcription. It regulates differentiation and survival during hemopoiesis. TAL-1 is also implicated in the genesis of hemopoietic maligcies and it may play an important role in hemopoietic differentiation. It serves as a positive regulator of eryhtroid differentiation. Mutations are associated with T-cell leukemia and melanoma. TAL-1 is know to bind to the LIM domain containing protein Rhombotin-2.Specifications
| TAL1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17542 | |
| TAL1 | |
| Full length human recombit protein of human TAL1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI6H6 | |
| Unconjugated | |
| TAL1 | |
| SCL, TCL5, bHLHa17, tal-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6886 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction