missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAL1 Monoclonal Antibody (OTI6H6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503919S
This item is not returnable.
View return policy
Description
TAL1 Monoclonal antibody specifically detects TAL1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| TAL1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6IQ20 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 222236 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI6H6 | |
| Unconjugated | |
| NAPEPLD | |
| basic helix-loop-helix transcription factor; bHLHa17; Class A basic helix-loop-helix protein 17; HGNC:11556; LOW QUALITY PROTEIN: T-cell acute lymphocytic leukemia protein 1; Scl; scl/tal1; SCL/tal-1; SCL-alpha; SCL-beta; si:ch211-135j1.3; stem cell leukemia protein; stem cell leukemia protein SCL; stem cell protein; stem-cell leukemia hematopoietic transcription factor; T cell acute lymphocytic leukemia 1; TAL bHLH transcription factor 1, erythroid differentiation factor; TAL1; Tal-1; tal-1 product; T-cell acute lymphocytic leukemia 1; T-cell acute lymphocytic leukemia protein 1; T-cell acute lymphocytic leukemia protein 1 homolog; T-cell leukemia/lymphoma protein 5; TCL5; transcription factor; zgc:85605; zSCL | |
| Full length human recombinant protein of human NAPEPLD produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction