missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talabostat isomer mesylate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Talabostat isomer mesylate is an isomer of talabostat mesylate. Talabostat (PT100, Val-boroPro) is a potent, nonselective and orally available dipeptidyl peptidase IV (DPP-IV) inhibitor with a Ki of 0.18 nM.
Specifications
Specifications
| Chemical Name or Material | Talabostat isomer mesylate |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C10H23BN2O6S |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (322.39 mM; Need ultrasonic) |
| SMILES | OB([C@@H]1N(C([C@H](N)C(C)C)=O)CCC1)O.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 310.18 |
| Formula Weight | 310.18 |
| Percent Purity | 98.0% |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction