missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TALDO1 Monoclonal Antibody (OTI2E4)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527220
This item is not returnable.
View return policy
Description
TALDO1 Monoclonal Antibody for Western Blot, IHC (P)
Transaldolase 1 is a key enzyme of the nonoxidative pentose phosphate pathway providing ribose-5-phosphate for nucleic acid synthesis and NADPH for lipid biosynthesis. This pathway can also maintain glutathione at a reduced state and thus protect sulfhydryl groups and cellular integrity from oxygen radicals. The functional gene of transaldolase 1 is located on chromosome 11 and a pseudogene is identified on chromosome 1 but there are conflicting map locations. The second and third exon of this gene were developed by insertion of a retrotransposable element. This gene is thought to be involved in multiple sclerosis.
Specifications
| TALDO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P37837 | |
| TALDO1 | |
| Full length human recombinant protein of TALDO1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E4 | |
| Unconjugated | |
| TALDO1 | |
| dihydroxyacetone transferase; glycerone transferase; TAL; TALDO; TALDO1; TALDOR; TALH; TAL-H; testicular secretory protein Li 56; transaldolase; transaldolase 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 6888 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction