missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Talin Monoclonal Antibody (8D4)

Description
The antibody detects a 240 kDa* protein corresponding to the molecular mass of Talin on SDS-PAGE immunoblots of human A431, rat PC12, and rabbit fibroblast cells.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
Specifications
Specifications
| Antigen | Talin |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8D4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | TLN2 |
| Gene Accession No. | P26039, P54939, Q71LX4, Q9Y490, Q9Y4G6 |
| Gene Alias | 5730421P04Rik; AI507121; AI787438; AL118320; ILWEQ; KIAA0320; KIAA1027; mKIAA0320; RGD1565416; talin 1; talin 2; talin-1; Talin-2; TLN; Tln1; Tln2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?