missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin1 Antibody (93E12), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50319AF750
This item is not returnable.
View return policy
Description
Talin1 Monoclonal specifically detects Talin1 in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Talin1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| TLN1 | |
| Recombinant protein fragment | |
| 0.1 mL | |
| 7094 | |
| Mouse | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 93000000000000 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ILWEQ, KIAA1027, talin 1, talin-1, TLN | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Does not cross react with Talin2. | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction