missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Talin2 Antibody (68E7), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50322V
This item is not returnable.
View return policy
Description
Talin2 Monoclonal specifically detects Talin2 in Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Talin2 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TLN2 | |
| recombinant protein fragment | |
| 0.1 mL | |
| Cytoskeleton Markers | |
| 83660 | |
| Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 680000000 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| DKFZp451B1011, DKFZp686I0976, DKFZp686K0979, ILWEQ, KIAA0320, talin2, talin-2, TLN 2, TLN2 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Does not cross react with Talin1. | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction