missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Taltobulin

Product Code. 16432737
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16432737

Brand: MedChemExpress HY15584

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Taltobulin (HTI-286), a synthetic analogue of the tripeptide hemiasterlin, is a potent antimicrotubule agent that circumvents P-glycoprotein-mediated resistance in vitro and in vivo. Taltobulin inhibits the polymerization of purified tubulin, disrupts microtubule organization in cells, and induces mitotic arrest, as well as apoptosis.

Specifications

Purity 98.07%
CAS 228266-40-8
Content And Storage Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 473.65 g/mol
Product Type Reference compound
Quantity 50 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (211.13 mM)
Synonym HTI-286∣ SPA-110
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.