missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAMRA Antibody (5G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22472
This item is not returnable.
View return policy
Description
TAMRA Monoclonal specifically detects TAMRA in Non-species specific samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| TAMRA | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunoprecipitation 10 ug | |
| Mouse | |
| Protein G purified | |
| RUO | |
| This antibody reacts with Chemical. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunoprecipitation | |
| 5G5 | |
| Unconjugated | |
| BP-25606, F07-0040, TAMRA Azide, TAMRA Azide, isomer 5, tetramethylrhodamine azide | |
| TAMRA molecule | |
| 100 μg | |
| Primary | |
| Non-species specific | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction