Learn More
Invitrogen™ Tamrintamab Chimeric Recombinant Human Monoclonal Antibody
GREENER_CHOICE
Human Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542238
Product Code. 17708863
  • € 524.00 / 100µg
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Tamrintamab is a biosimilar that targets DPEP3. This gene encodes a membrane-bound glycoprotein from the family of dipeptidases involved in hydrolytic metabolism of various dipeptides, including penem and carbapenem beta-lactam antibiotics. This gene is located on chromosome 16 in a cluster with another member of this family. Alternatively spliced transcript variants that encode different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tamrintamab Chimeric
Recombinant Monoclonal
Unconjugated
SC-003 mAb; SC34.28ss1; SC-Mab003
Human DPEP3/MBD3.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical
ELISA
5 mg/mL
PBS with no preservative; pH 7.4
Human
Protein A
RUO
Human
Antibody
IgG1 κ