Learn More
BOLA Tandem Magnetic Stirring Bars
Product Code. 10273452
Click to view available options
Diameter (Metric):
40 mm
55 mm
110 mm
155 mm
Unit Size:
Each
Product Code. 10273452
Brand: BOLA C36326
Only null left
Add to basket
Add to basket
Tandem Magnetic Stirring Bars
  • PTFE-encapsulated magnetic cores (Alnico 5), center bore for receiving the BOLA Bearing Neck or a glass neck (available from a glassblower), universal chemical resistance.
  • Bearing neck not included in delivery.
Specifications
Autoclavable Autoclavable
Color White
Dimensions (L x W x H) 34 x 14 x 14 mm
Coating PTFE
Diameter (Metric) 40 mm