Learn More
BOLA Tandem Magnetic Stirring Bars
Click to view available options
Diameter (Metric):
40 mm
55 mm
110 mm
155 mm
Unit Size:
Each
Description
- PTFE-encapsulated magnetic cores (Alnico 5), center bore for receiving the BOLA Bearing Neck or a glass neck (available from a glassblower), universal chemical resistance.
- Bearing neck not included in delivery.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Dimensions (L x W x H) | 34 x 14 x 14 mm |
| Coating | PTFE |
| Diameter (Metric) | 40 mm |