missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tandutinib, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Description
Tandutinib (MLN518) is a potent and selective inhibitor of the FLT3 with an IC50 of 0.22 μM, and also inhibits c-Kit and PDGFR with IC50s of 0.17 μM and 0.20 μM, respectively. Tandutinib can be used for acute myelogenous leukemia (AML). Tandutinib has the ability to cross the blood-brain barrier.
Specifications
Specifications
| Chemical Name or Material | Tandutinib |
| CAS | 387867-13-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C31H42N6O4 |
| Quantity | 100 mg |
| Synonym | MLN518 CT53518 |
| Solubility Information | DMSO : 50 mg/mL (88.86 mM; Need ultrasonic) |
| SMILES | O=C(N1CCN(C2=C(C(C=C3OCCCN4CCCCC4)=NC=N2)C=C3OC)CC1)NC5=CC=C(OC(C)C)C=C5 |
| Molecular Weight (g/mol) | 562.7 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction