Learn More
Novus Biologicals™ Tangential Flow Filtration (TFF)-EV Concentrator
Shop All Bio Techne Products
Click to view available options
Quantity:
1 Units
5 Units
Unit Size:
Each
Pack of 5
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Quantity | 5 Units |
| Sample Type | Diluted matrices (urine and cell culture supernatant) |