Learn More
Invitrogen™ TANGO2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA569912
Product Code. 15828323
  • € 530.00 / 100µL
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
This gene belongs to the transport and Golgi organization family, whose members are predicted to play roles in secretory protein loading in the endoplasmic reticulum. Depletion of this gene in Drosophila S2 cells causes fusion of the Golgi with the ER. In mouse tissue culture cells, this protein co-localizes with a mitochondrially targeted mCherry protein and displays very low levels of co-localization with Golgi and peroxisomes. Allelic variants of this gene are associated with rhabdomyolysis, metabolic crises with encephalopathy, and cardiac arrhythmia.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TANGO2
Polyclonal
Unconjugated
TANGO2
C17H22orf25; C22orf25; C26H22orf25; D16H22S680E; DKFZP761P1121 protein; hypothetical protein DKFZp761P1121; MECRCN; RGD1310348; ser/Thr-rich protein T10 in DGCR region; T10; TANGO2; transport and golgi organization 2; transport and golgi organization 2 homolog; transport and golgi organization 2 homolog (Drosophila); transport and Golgi organization protein 2 homolog; Uncharacterized protein C22orf25; uncharacterized protein C22orf25 homolog; wu:fj48d05; zgc:100952
Rabbit
Affinity chromatography
RUO
128989
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q6ICL3
TANGO2
synthetic peptide directed towards the N terminal region of human TANGO2.