Learn More
TANK Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27065-1-AP-20UL
 View more versions of this product
Product Code. 16865355
  • € 145.00 / 20µL
Description
TANK was initially identified as a novel TRAF-interacting protein that regulated TRAF-mediated signal transduction. Specifically, ligand binding by surface receptors in the tumor necrosis factor (TNF) receptor and Toll/interleukin-1 (IL-1) receptor families lead to the formation of a TRAF/TANK complex that mediates the activation of the transcription factor NF-kappa-B. This activation of NF-kappa-B occurs through an association with the kinases IKK-iota and TBK1. More recently, it was shown that these proteins can then form a complex with NEMO, a protein that regulates the activity of the Ikappa-B complex. This suggests that in addition to the possibility that TBK1 and IKK-iota activate the IKKs, the association with the IKK complex may help these kinases modulate other functions, such as the transactivation potential of NF-kappa-B proteins. At least two isoforms of TANK are known to exist.
Specifications
Specifications
TANK
Polyclonal
Unconjugated
TANK
I TRAF, ITRAF, TANK, TRAF interacting protein, TRAF2
Rabbit
Antigen Affinity Chromatography
RUO
10010, 21353
-20°C
Liquid
Western Blot
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P70347, Q92844
Tank
TANK Fusion Protein Ag25740
20 μL
Primary
Human, Mouse