Learn More
TAO Kinase 1 Antibody (TAO1 1.2), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50324B
Product Code. 18114558
  • € 437.00 / 0.10mL
Description
TAO Kinase 1 Monoclonal specifically detects TAO Kinase 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TAO Kinase 1
Monoclonal
Biotin
PBS with 0.05% Sodium Azide
TAOK1
His-tagged recombinant human TAO Kinase 1 (722-1001) generated in E.coli
0.1 mL
GPCR, Protein Kinase
57551
Store at 4C in the dark.
IgG2b
Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown
TAO1 1.2
Western Blot, Immunocytochemistry/Immunofluorescence
EC 2.7.11, FLJ14314, hKFC-B, KIAA1361STE20-like kinase PSK2, Kinase from chicken homolog B, MAP3K16EC 2.7.11.1, MARKKmicrotubule affinity regulating kinase kinase, PSK2serine/threonine kinase TAO1, serine/threonine-protein kinase TAO1, TAO kinase 1, TAO1serine/threonine protein kinase TAO1 homolog, Thousand and one amino acid protein 1
Mouse
Protein A purified