missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAO Kinase 1 Antibody (TAO1 1.2), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50324G
This item is not returnable.
View return policy
Description
TAO Kinase 1 Monoclonal specifically detects TAO Kinase 1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
| TAO Kinase 1 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TAOK1 | |
| His-tagged recombinant human TAO Kinase 1 (722-1001) generated in E.coli | |
| 0.1 mL | |
| GPCR, Protein Kinase | |
| 57551 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence, KnockDown | |
| TAO1 1.2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| EC 2.7.11, FLJ14314, hKFC-B, KIAA1361STE20-like kinase PSK2, Kinase from chicken homolog B, MAP3K16EC 2.7.11.1, MARKKmicrotubule affinity regulating kinase kinase, PSK2serine/threonine kinase TAO1, serine/threonine-protein kinase TAO1, TAO kinase 1, TAO1serine/threonine protein kinase TAO1 homolog, Thousand and one amino acid protein 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction