missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAP1, Mouse anti-Human, Clone: 2B4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006890-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: GSETRRLSLFLVLVVLSSLGEMAIPFFTGRLTDWILQDGSADTFTRNLTLMSILTIASAVLEFVGDGIYNNTMGHVHSHLQGEVFGAVLRQETEFFQQNQTGNIMSRVTESpecifications
| TAP1 | |
| Monoclonal | |
| Unconjugated | |
| transporter 1, ATP-binding cassette, sub-family B (MDR/TAP) | |
| ABC17/ABCB2/APT1/D6S114E/FLJ26666/FLJ41500/PSF1/RING4/TAP1*0102N/TAP1N | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2B4 | |
| In 1x PBS, pH 7.4 | |
| BC014081 | |
| TAP1 | |
| TAP1 (AAH14081, 241 a.a. ∼ 350 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6890 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction